Merck
  • Mechanical Stretching-Induced Traumatic Brain Injury Is Mediated by the Formation of GSK-3β-Tau Complex to Impair Insulin Signaling Transduction.
Mechanical Stretching-Induced Traumatic Brain Injury Is Mediated by the Formation of GSK-3β-Tau Complex to Impair Insulin Signaling Transduction.
Biomedicines (2021-11-28)
Pei-Wen Cheng, Yi-Chung Wu, Tzyy-Yue Wong, Gwo-Ching Sun, Ching-Jiunn Tseng
RÉSUMÉ
Traumatic brain injury confers a significant and growing public health burden. It is a major environmental risk factor for dementia. Nonetheless, the mechanism by which primary mechanical injury leads to neurodegeneration and an increased risk of dementia-related diseases is unclear. Thus, we aimed to investigate the effect of stretching on SH-SY5Y neuroblastoma cells that proliferate in vitro. These cells retain the dopamine-β-hydroxylase activity, thus being suitable for neuromechanistic studies. SH-SY5Y cells were cultured on stretchable membranes. The culture conditions contained two groups, namely non-stretched (control) and stretched. They were subjected to cyclic stretching (6 and 24 h) and 25% elongation at 1 Hz. Following stretching at 25% and 1 Hz for 6 h, the mechanical injury changed the mitochondrial membrane potential and triggered oxidative DNA damage at 24 h. Stretching decreased the level of brain-derived neurotrophic factors and increased amyloid-β, thus indicating neuronal stress. Moreover, the mechanical injury downregulated the insulin pathway and upregulated glycogen synthase kinase 3β (GSK-3β)S9/p-Tau protein levels, which caused a neuronal injury. Following 6 and 24 h of stretching, GSK-3βS9 was directly bound to p-TauS396. In contrast, the neuronal injury was improved using GSK-3β inhibitor TWS119, which downregulated amyloid-β/p-Taus396 phosphorylation by enhancing ERK1/2T202/Y204 and AktS473 phosphorylation. Our findings imply that the neurons were under stress and that the inactivation of the GSK3β could alleviate this defect.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-phospho-GSK3β (Ser9) Antibody, clone 2D3, clone 2D3, Upstate®, from mouse