Merck
  • Stomatin‑like protein 2 induces metastasis by regulating the expression of a rate‑limiting enzyme of the hexosamine biosynthetic pathway in pancreatic cancer.
Stomatin‑like protein 2 induces metastasis by regulating the expression of a rate‑limiting enzyme of the hexosamine biosynthetic pathway in pancreatic cancer.
Oncology reports (2021-04-14)
Dang Chao, Kyohei Ariake, Satoko Sato, Hideo Ohtsuka, Tatsuyuki Takadate, Masaharu Ishida, Kunihiro Masuda, Shimpei Maeda, Takayuki Miura, Katsutaka Mitachi, Xun Jing Yu, Fumiyoshi Fujishima, Masamichi Mizuma, Kei Nakagawa, Takanori Morikawa, Takashi Kamei, Michiaki Unno
RÉSUMÉ
Stomatin‑like protein 2 (SLP‑2) is associated with poor prognosis in several types of cancer, including pancreatic cancer (PC); however, the molecular mechanism of its involvement remains elusive. The present study aimed to elucidate the role of this protein in the development of PC. Human PC cell lines AsPC‑1 and PANC‑1 were transfected by a vector expressing SLP‑2 shRNA. Analyses of cell proliferation, migration, invasion, chemosensitivity, and glucose uptake were conducted, while a mouse xenograft model was used to evaluate the functional role of SLP‑2 in PC. Immunohistochemical analysis was retrospectively performed on human tissue samples to compare expression between the primary site (n=279) and the liver metastatic site (n=22). Furthermore, microarray analysis was conducted to identify the genes correlated with SLP‑2. In vitro analysis demonstrated that cells in which SLP‑2 was suppressed exhibited reduced cell motility and glucose uptake, while in vivo analysis revealed a marked decrease in the number of liver metastases. Immunohistochemistry revealed that SLP‑2 was increased in liver metastatic sites. Microarray analysis indicated that this protein regulated the expression of glutamine‑fructose‑6‑phosphate transaminase 2 (GFPT2), a rate‑limiting enzyme of the hexosamine biosynthesis pathway. SLP‑2 contributed to the malignant character of PC by inducing liver metastasis. Cell motility and glucose uptake may be induced via the hexosamine biosynthesis pathway through the expression of GFPT2. The present study revealed a new mechanism of liver metastasis and indicated that SLP‑2 and its downstream pathway could provide novel therapeutic targets for PC.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Conjugué anticorps anti-IgG de lapin (molécule entière)-peroxydase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Test d'invasion cellulaire QCM ECMatrix, 24 puits (8 µm), fluorimétrique, The Cell Invasion Assay Kit kuses a 24 well plate with 8 um pores, which is ideal for evaluation of invasive tumor cells.
Sigma-Aldrich
Essai de chimiotaxie/migration cellulaire QCM, 24 puits (8 µm), fluorimétrique, The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with fluorescent detection.