Merck
High Prevalence of Rare Monogenic Forms of Obesity in Obese Guadeloupean Afro-Caribbean Children.
The Journal of clinical endocrinology and metabolism (2017-12-08)
Lydia Foucan, Laurent Larifla, Emmanuelle Durand, Christine Rambhojan, Christophe Armand, Carl-Thony Michel, Rachel Billy, Véronique Dhennin, Franck De Graeve, Iandry Rabearivelo, Olivier Sand, Jean-Marc Lacorte, Philippe Froguel, Amélie Bonnefond
RÉSUMÉ
The population of Guadeloupe Island exhibits a high prevalence of obesity. We aimed to investigate whether rare genetic mutations in genes involved in monogenic obesity (or diabetes) might be causal in this population of Afro-Caribbean ancestry. This was a secondary analysis of a study on obesity conducted in schoolchildren from Guadeloupe in 2013 that aimed to assess changes in children's profiles after a lifestyle intervention program. Through next-generation sequencing, we sequenced coding regions of 59 genes involved in monogenic obesity or diabetes in participants from this study. A total of 25 obese schoolchildren from Guadeloupe were screened for rare mutations (nonsynonymous, splice-site, or insertion/deletion) in 59 genes. Correlation between phenotypes and mutations of interest. We detected five rare heterozygous mutations in five different children with obesity: MC4R p.Ile301Thr and SIM1 p.Val326Thrfs*43 mutations that were pathogenic; SIM1 p.Ser343Pro and SH2B1 p.Pro90His mutations that were likely pathogenic; and NTRK2 p.Leu140Phe that was of uncertain significance. In parallel, we identified seven carriers of mutations in ABCC8 (p.Lys1521Asn and p.Ala625Val) or KCNJ11 (p.Val13Met and p.Val151Met) that were of uncertain significance. We were able to detect pathogenic or likely pathogenic mutations linked to severe obesity in >15% of this population, which is much higher than what we observed in Europeans (∼5%).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Millipore
Test ELISA "Dual Range" de la leptine humaine, This Human Leptin "Dual Range" ELISA is used to measure & quantify Leptin levels in Metabolism & Endocrine & Neuroscience research.