Merck
  • Sphingosine 1-phosphate (S1P) regulates vascular contraction via S1P3 receptor: investigation based on a new S1P3 receptor antagonist.
Sphingosine 1-phosphate (S1P) regulates vascular contraction via S1P3 receptor: investigation based on a new S1P3 receptor antagonist.
Molecular pharmacology (2010-01-26)
Akira Murakami, Hiroshi Takasugi, Shinya Ohnuma, Yuuki Koide, Atsuko Sakurai, Satoshi Takeda, Takeshi Hasegawa, Jun Sasamori, Takashi Konno, Kenji Hayashi, Yoshiaki Watanabe, Koji Mori, Yoshimichi Sato, Atsuo Takahashi, Naoki Mochizuki, Nobuyuki Takakura
RÉSUMÉ
Sphingosine 1-phosphate (S1P) induces diverse biological responses in various tissues by activating specific G protein-coupled receptors (S1P(1)-S1P(5) receptors). The biological signaling regulated by S1P(3) receptor has not been fully elucidated because of the lack of an S1P(3) receptor-specific antagonist or agonist. We developed a novel S1P(3) receptor antagonist, 1-(4-chlorophenylhydrazono)-1-(4-chlorophenylamino)-3,3-dimethyl- 2-butanone (TY-52156), and show here that the S1P-induced decrease in coronary flow (CF) is mediated by the S1P(3) receptor. In functional studies, TY-52156 showed submicromolar potency and a high degree of selectivity for S1P(3) receptor. TY-52156, but not an S1P(1) receptor antagonist [(R)-phosphoric acid mono-[2-amino-2-(3-octyl-phenylcarbamoyl)-ethyl] ester; VPC23019] or S1P(2) receptor antagonist [1-[1,3-dimethyl-4-(2-methylethyl)-1H-pyrazolo[3,4-b]pyridin-6-yl]-4-(3,5-dichloro-4-pyridinyl)-semicarbazide; JTE013], inhibited the decrease in CF induced by S1P in isolated perfused rat hearts. We further investigated the effect of TY-52156 on both the S1P-induced increase in intracellular calcium ([Ca(2+)](i)) and Rho activation that are responsible for the contraction of human coronary artery smooth muscle cells. TY-52156 inhibited both the S1P-induced increase in [Ca(2+)](i) and Rho activation. In contrast, VPC23019 and JTE013 inhibited only the increase in [Ca(2+)](i) and Rho activation, respectively. We further confirmed that TY-52156 inhibited FTY-720-induced S1P(3) receptor-mediated bradycardia in vivo. These results clearly show that TY-52156 is both sensitive and useful as an S1P(3) receptor-specific antagonist and reveal that S1P induces vasoconstriction by directly activating S1P(3) receptor and through a subsequent increase in [Ca(2+)](i) and Rho activation in vascular smooth muscle cells.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
TY-52156, ≥98% (HPLC)