Merck
The EcoA restriction and modification system of Escherichia coli 15T-: enzyme structure and DNA recognition sequence.
The EMBO journal (1984-03-01)
B Suri, J C Shepherd, T A Bickle
RÉSUMÉ
The EcoA restriction enzyme from Escherichia coli 15T- has been isolated. It proves to be an unusual enzyme, clearly related functionally to the classical type I restriction enzymes. The basic enzyme is a two subunit modification methylase. Another protein species can be purified which by itself has no enzymatic activities but which converts the modification methylase to an ATP and S-adenosylmethionine-dependent restriction endonuclease. The DNA recognition sequence of EcoA has an overall structure that is very similar to previously determined type I sequences. It is: 5'-GAGNNNNNNNGTCA-3' 3'-CTCNNNNNNNCAGT-5' where N can be any nucleotide. Modification methylates the adenosyl residue in the specific trinucleotide and the adenosyl residue in the lower strand of the specific tetranucleotide.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Adenosine 5′-triphosphate–Agarose, aqueous glycerol suspension