Merck
Near-infrared-triggered photon upconversion tuning in all-inorganic cesium lead halide perovskite quantum dots.
Nature communications (2018-08-29)
Wei Zheng, Ping Huang, Zhongliang Gong, Datao Tu, Jin Xu, Qilin Zou, Renfu Li, Wenwu You, Jean-Claude G Bünzli, Xueyuan Chen
RÉSUMÉ
All-inorganic CsPbX3 (X = Cl, Br, and I) perovskite quantum dots (PeQDs) have shown great promise in optoelectronics and photovoltaics owing to their outstanding linear optical properties; however, nonlinear upconversion is limited by the small cross-section of multiphoton absorption, necessitating high power density excitation. Herein, we report a convenient and versatile strategy to fine tuning the upconversion luminescence in CsPbX3 PeQDs through sensitization by lanthanide-doped nanoparticles. Full-color emission with wavelengths beyond the availability of lanthanides is achieved through tailoring of the PeQDs bandgap, in parallel with the inherent high conversion efficiency of energy transfer upconversion under low power density excitation. Importantly, the luminescent lifetimes of the excitons can be enormously lengthened from the intrinsic nanosecond scale to milliseconds depending on the lifetimes of lanthanide ions. These findings provide a general approach to stimulate photon upconversion in PeQDs, thereby opening up a new avenue for exploring novel and versatile applications of PeQDs.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Octadecene, technical grade, 90%