Merck
PARP3 is a promoter of chromosomal rearrangements and limits G4 DNA.
Nature communications (2017-04-28)
Tovah A Day, Jacob V Layer, J Patrick Cleary, Srijoy Guha, Kristen E Stevenson, Trevor Tivey, Sunhee Kim, Anna C Schinzel, Francesca Izzo, John Doench, David E Root, William C Hahn, Brendan D Price, David M Weinstock
RÉSUMÉ
Chromosomal rearrangements are essential events in the pathogenesis of both malignant and nonmalignant disorders, yet the factors affecting their formation are incompletely understood. Here we develop a zinc-finger nuclease translocation reporter and screen for factors that modulate rearrangements in human cells. We identify UBC9 and RAD50 as suppressors and 53BP1, DDB1 and poly(ADP)ribose polymerase 3 (PARP3) as promoters of chromosomal rearrangements across human cell types. We focus on PARP3 as it is dispensable for murine viability and has druggable catalytic activity. We find that PARP3 regulates G quadruplex (G4) DNA in response to DNA damage, which suppresses repair by nonhomologous end-joining and homologous recombination. Chemical stabilization of G4 DNA in PARP3-/- cells leads to widespread DNA double-strand breaks and synthetic lethality. We propose a model in which PARP3 suppresses G4 DNA and facilitates DNA repair by multiple pathways.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-phospho-histone H2A.X (Ser139), clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
PhenDC3, ≥97% (HPLC)