Principaux documents
K5130-U
Omnifit® EZ Benchmark Columns with 2 Fixed Endpieces
I.D. × L 6.6 mm × 400 mm
1 EA
340,00 €
340,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Changer de vue
1 EA
340,00 €
About This Item
Code UNSPSC :
23151816
Nomenclature NACRES :
SB.52
340,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Fabricant/nom de marque
Omnifit 006EZ-06-40-FF
Niveau de qualité
Technique(s)
LPLC: suitable
D.I. × L
6.6 mm × 400 mm
Description générale
Omnifit® Labware complete columns are supplied assembled and ready for use, with tubing and fittings to connect the column into an existing system. Two configurations are available:
  • Non-adjustable with two fixed endpieces (-FF), a cost-effective option when height adjustment is not important.
  • One fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80mm of height adjustment.
Informations légales
Omnifit is a registered trademark of Diba Industries