Principaux documents
2631725
Supelco
Inlet Liner, for Packed GC (wool packed)
wool packed, pkg of 25 ea
About This Item
Code UNSPSC :
41121809
Conditionnement
pkg of 25 ea
L × D.E. × D.I.
112 mm × 6.0 mm × 3.0 mm
Description générale
Deactivated (silanized) glass inlet liner for packed injection ports.
  • Fits PerkinElmer (AutoSystem)