Principaux documents
23315
Supelco
Dispositif anti-retour d′hydrogène
1 EA
294,00 €
294,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Changer de vue
1 EA
294,00 €
About This Item
Code UNSPSC :
41112500
294,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Description générale
Install a flash arrestor downstream of a hydrogen gas cylinder (see figure), or a hydrogen generator. In event of a flashback, the flash arrestor diverts the flame into three feet (1 m) of tubing, where the flame is extinguished and the heat is absorbed. The shock wave preceding the flashback closes and locks the arrestor′s shutoff valve, eliminating the flow of gas. Inlet/outlet fittings are 1/4 in. female NPT. For use to 50 psig (3.5 kg/cm2). Meets Occupational Safety and Health Administration (OSHA) and National Fire Protection Agency (NFPA) codes. Factory Mutual approved.