Principaux documents
Z762806
Eppendorf® Combitips advanced® pipette tips, Biopur® grade
maximum volume 10 mL
Synonyme(s) :
pipetting tips
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
orange tip
Stérilité
sterile
Caractéristiques
barrier: no
Conditionnement
pack of 100 ea individually wrapped
Fabricant/nom de marque
Eppendorf® 0030089677
Volume maximum
10 mL
Description générale
The Combitips advanced are optimized for all Repeater models to create the perfect connection!
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
The "Biopur" tips are individually blister-wrapped and feature an access tab which makes them easier to open, even with gloves.
Caractéristiques et avantages
  • pyrogen-free
  • free of human DNA
  • free of bacterial DNA
  • RNase-free
  • PCR inhibitor-free
  • DNase-free
  • ATP-free
Autres remarques
Browse all pipette tips
Informations légales
Biopur is a registered trademark of Eppendorf AG
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG