Principaux documents
Z744144
Wilmad® 50 ml Suprasil® Small Volume Liquid Nitrogen Dewar
pack of 1
Synonyme(s) :
EPR cryo dewar, EPR dewar
About This Item
Code UNSPSC :
41121804
Conditionnement
pack of 1
Fabricant/nom de marque
Wilmad WG-816-Q
Description générale
Dewar Flasks provide a convenient alternative to Variable Temperature systems for studies at 77 K, and are silvered around the reservoir.
Informations légales
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries