Z744065
EZcap™ PCR FilmStrips™
polyester, non-sterile
About This Item
Code UNSPSC :
41122410
Nomenclature NACRES :
NB.04
Produits recommandés
Matériaux
polyester
Stérilité
non-sterile
Conditionnement
case of 400 ea
Fabricant/nom de marque
Excel Scientific US-CS1X8-400
Technique(s)
PCR: suitable
Catégories apparentées
Description générale
- An economical and more user-friendly alternative to strip-caps; no more sore thumbs and fingers guaranteed!
- Precision cut to a smaller dimension to seal one 8-well row of a 96-well plate or a single PCR 8-tube strip
- Each sheet contains 8 FilmStrips on a continuous backing liner for ease of use
- Strips can be peeled individually or two-at-a-time for increased efficiency
- Strips can be used to selectively cover one row of a plate to prevent accidental pipetting or to re-seal rows that have been pierced
- Certified DNase- & RNase- free
- 2.0 mil clear polyester film
- Specially formulated adhesive provides an excellent seal to minimize evaporation, reduce cross-contamination and prevent spillage
- FilmStrips remove easily and cleanly with no splash back as can occur with strip-caps
- Features two removable end-tabs with perforations for ease of handling, application and removal
- A non-locking workstation is recommended for sample prep (e.g. 96 round-well PCR plate or deep well block)
- Removal of the end-tabs at the perforations is recommended before cycling
- A final sealing step is highly recommended in the thermal cycler because 8-tube strips flex during handling
Informations légales
EZcap is a trademark of Excel Scientific, Inc.
FilmStrips is a trademark of Excel Scientific, Inc.