Principaux documents
Z741028
Nalgene® media bottles, style 2020
capacity 2000 mL
Synonyme(s) :
bottle nalgene, nalgene containers, sterile bottles
12 EA
348,00 €
348,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
12 EA
348,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
348,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
high-density polyethylene cap
square bottle
transparent PETG
Joint
Joints threaded neck (53B)
Description
Not autoclavable
Stérilité
sterile; γ-irradiated
Caractéristiques
graduations: no
with cap
Conditionnement
pack of 12 ea (2 xpack of 6)
Fabricant/nom de marque
Nalgene® 342020-2000
Capacité
2000 mL
Catégories apparentées
Comparer avec des articles similaires
Voir la comparaison complète
Montrer les différences
1 of 4
Cet article
Z364541Z364568Z364576
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
capacity
2000 mL
capacity
125 mL
capacity
250 mL
capacity
500 mL
feature
graduations: no, with cap
feature
graduations: no, with cap
feature
graduations: no, with cap
feature
graduations: no, with cap
joint
Joints threaded neck (53B)
joint
Joints threaded neck (38-430mm)
joint
Joints threaded neck (38-430mm)
joint
Joints threaded neck (38-430mm)
manufacturer/tradename
Nalgene® 342020-2000
manufacturer/tradename
Nalgene® 342020-0125
manufacturer/tradename
Nalgene® 342020-0250
manufacturer/tradename
Nalgene® 342020-0500
Description générale
Media bottles (square). Same materials and design as Style 2019, but without shrink band around neck.
Caractéristiques et avantages
  • Extensive validation documentation is available
  • Produced from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG) that enhance reduced permeability to CO2/O2.
  • Tamper evident shrink sleeves used along with moulded in graduations and neck ring.
  • Non-pyrogenic and non-cytotoxic
  • Animal derived component free (ADCF) material for bottle and closure material.
  • Leakproof and transparent
  • Devoid of costly washing, depyrogenation steps
  • Prolonged storage of pH-sensitive liquids such as culture media
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries