Principaux documents
Z737836
Jenco stereo microscopes, dissecting
AC/DC input 220 V AC, binocular 10x-20x, illumination: halogen top; fluorescent bottom
Synonyme(s) :
dissecting microscopes, microscopes, stereo microscopes
About This Item
Code UNSPSC :
41191506
Nomenclature NACRES :
MA.44
Description
3 Plug
EU 2-prong plug
Fabricant/nom de marque
Jenco ST-F801-2
Description générale
Dissection and observation of macro specimens is more enlightening with a Jenco Stereo microscope. The large stage allows ample room for sample manipulation but still fits in a storage cabinet.
Their illuminated stands let you choose light from three directions - above, below or both, to help observe opaque or transparent samples. Solid, all metal focusing mechanisms will endure student use well beyond the five year warranty.
Teachers like them because the eyepieces are locked and don′t walk, the all metal frames are built to last and the low voltage bulbs last for hundreds of hours and are easy to replace. Even models with rechargeable illuminators are available.
  • Affordable
  • Portable
  • Durable
  • Easy to use
  • Five Year warranty
Informations légales
Jenco is a trademark of Jenco International, Inc.