Z719943
Microcentrifuge Tube Opener
double-ended tool opens both small and larger tubes
About This Item
Code UNSPSC :
41121702
Nomenclature NACRES :
NB.11
Produits recommandés
Fabricant/nom de marque
Bel-Art
Technique(s)
cryopreservation: suitable
Description générale
Easily open microcentrifuge tubes with this double-ended tool. Works for all sizes of tubes. One end opens standard 1.5, 2 and 2.2 mL tubes while the other end has a forked blade that opens smaller sizes (250, 400 and 550 μL tubes). 79mm (31/8 in.) long.