Principaux documents
Z692727
microDOC Compact gel documentation system, with UV Transilluminator (UVT254)
with TotalLab 1D software, AC/DC input 230 V AC
About This Item
Code UNSPSC :
41105300
Nomenclature NACRES :
NB.22
Caractéristiques
Contains UK and Schuko plugs
Fabricant/nom de marque
Cleaver CSI-MDOCUV2541D
Description générale
This option combines the microDOC Compact Gel Documentation System with a 12 x 12 cm UV 254 nm tranilluminator.
Available with optional TotalLab 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
12 x 12 cm UV 254 nm transilluminator
Informations légales
TotalLab is a trademark of Nonlinear Dynamics Ltd.