Documents
Z659223
Bandelin SONOPULS ultrasonic homogenizer accessories
Stand HG 5
Synonyme(s) :
transducer support frame
About This Item
Code UNSPSC :
41122800
Matériaux
stainless steel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bandelin 459
Disponibilité
available only in EU
Description générale
With lab clamp, for safe fixing of ultrasonic converter