Principaux documents
Z646628
BRAND® Transferpette® S pipette, fixed volume, single channel
volume 25 μL, F-25
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Fabricant/nom de marque
BRAND 704720
Taille de la pointe
, 2-200 μL, 5-300μL & *1-100μL
Volume
25 μL
Description générale
For more than 25 years, BRAND has been developing and manufacturing piston-operated pipettes. The new Transferpette S pipettes are the product of this long experience, intensive ergonomics studies, and the use of innovative materials.
The Transferpette S pipettes are the perfect manual pipettes for the most demanding applications in the lab.
They include all of the features requested by Life Science users: rugged construction, complete autoclavability, highest precision, and Easy Calibration technology for convenient, long-term reliability.
  • Familiar centrally placed pipetting key and separate tip ejector
  • Ergonomic finger rest that adapts to your hand for a relaxed grip
  • Complete autoclavability at 121°C (2 bar), acc. to DIN EN 285
  • Color-coding for easy tip selection
  • Corrosion-resistant piston and ejector for durability
Includes: Transferpette S, Fixed volume, conformity certified, with performance certificate, wall/rack mount.
* Filter tips
Caractéristiques et avantages
• Efficient: one-hand operation• Flexible: ergonomic design for all hand sizes
Autres remarques
BR704720 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG