Z637122
BRAND® UV cuvette lids, PE
round yellow lid
About This Item
Matériaux
round yellow lid
Fabricant/nom de marque
BRAND
Description générale
Round covers allow a tight closure and make it possible to store samples down to -20°C.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG