Principaux documents
Z325104
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 100 mL, one mark
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
AR-Glas®
Description
total delivery Ex + 15 sec waiting time
Caractéristiques
class AS
Conditionnement
pack of 6 ea
Fabricant/nom de marque
BRAND 29719
Plage de mesure du volume
100 mL, accuracy: 0.08 mL
Capacité
100 mL
Description générale
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
Autres remarques
BR29719 (India)
Informations légales
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Protocoles
This page contains detailed information on working with and handling pipettes and pipetting aids.