Merck
Passer à
WPB01474
Whirl-Pak® Blender Bag
capacity 5441 mL (184 oz), bag W × L × thickness 37.5 cm × 50 cm × 0.102 mm
Synonyme(s) :
Nasco B01474, homogenizer sample bags, sample bag, sample receptacle, sample storage bag
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
Taille/RéférenceDisponibilitéPrix
A propos de cet article
NACRES:
NB.74
UNSPSC Code:
24111500
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
material
clear linear low-density polyethylene (LLDPE)
sterility
sterile
packaging
pkg of 500 ea (2 boxes of 250EA)
manufacturer/tradename
Whirl-Pak® B01474
bag W × L × thickness
37.5 cm × 50 cm × 0.102 mm
capacity
5441 mL (184 oz)
General description
These bags are ideal for use in a homogenizer in the lab with all solid and liquid contents including but not limited to: food, protein, powder, pharmaceuticals, dairy, environmental and many more. Whirl-Pak® sterilized sampling bags are ideal for food & beverage, dairy, water, medical, veterinary, environmental, soil, and industrial markets as well as surface, forensic science, genetics, biomedical, and pharmaceutical sampling. Whirl-Pak® bags have been the professionals’ trusted sampling bag for over 60 years and offer puncture-proof closure tabs, guaranteed sterility, easy identification, and a leak-proof closure you can rely on.
Features and Benefits
  • Perforated for easy opening and to protect sterility
  • Designed for quick testing and disposal when the sample does not have to be retained
  • Bag can be frozen to any temperature, even with liquid nitrogen, but should not be used at temperatures above 180°F (82°C)
  • FDA and USDA Approved
  • Post-manufacturing sterilization
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Comparer avec des articles similaires
Voir la comparaison complète
Montrer les différences
1 of 1
Cet article
WPB01421WPB01027WAWPB00990WA
description
capacity 5441 mL (184 oz), bag W × L × thickness 37.5 cm × 50 cm × 0.102 mm
description
capacity 1627  mL (55 oz), bag W × L × thickness 19 cm × 30 cm × 0.076 mm
description
1242 mL capacity(42 oz)
description
798 mL capacity(27 oz)
capacity
5441 mL (184 oz)
capacity
1627  mL (55 oz)
capacity
1242 mL capacity(42 oz)
capacity
798 mL capacity(27 oz)
material
clear linear low-density polyethylene (LLDPE)
material
clear linear low-density polyethylene (LLDPE)
material
-
material
-
bag W × L × thickness
37.5 cm × 50 cm × 0.102 mm
bag W × L × thickness
19 cm × 30 cm × 0.076 mm
bag W × L × thickness
-
bag W × L × thickness
-
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
manufacturer/tradename
Whirl-Pak® B01474
manufacturer/tradename
Whirl-Pak® B01421
manufacturer/tradename
Whirl-Pak® B01027WA
manufacturer/tradename
Whirl-Pak® B00990WA
packaging
pkg of 500 ea (2 boxes of 250EA)
packaging
pkg of 2000 ea (2 boxes of 1000EA)
packaging
pkg of 500 ea
packaging
pkg of 500 ea
Still not finding the right product?
Explore all of our products under Whirl-Pak® Blender Bag