Principaux documents
TMO382019-1000
Nalgene® Certified Clean bottles
clear PETG, volume 1000 mL, case of 24 ea
Synonyme(s) :
PETG bottle, certified nalgene bottle
24 EACH
607,00 €
607,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
24 EACH
607,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
607,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
clear PETG
Conditionnement
case of 24 ea
Fabricant/nom de marque
Nalgene 382019-1000
Capacité
1000 mL
Volume
1000 mL
Description générale
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific Nalgene PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries