Principaux documents
TMO374320-BR
Nunc® Low Profile Universal Tubes in 2D bar coded latch rack
polypropylene, capacity (5.0 mL), flat bottom, external thread, untreated, case of 10 × 24 ea
Synonyme(s) :
barcoded tube, sample storage tube
240 EACH
799,00 €
799,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
240 EACH
799,00 €
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
799,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
flat bottom
polypropylene
Conditionnement
case of 10 × 24 ea
Fabricant/nom de marque
Nunc 374320-BR
Capacité
(5.0 mL)
Catégories apparentées
Description générale
A robust solution for solid or liquid sample storage. Available in configurations that meet your tracking and packaging requirements, Thermo Scientific Nunc Low-Profile 5.0mL Externally-Threaded Universal Tubes feature 2D barcodes, graduations, a white patch for labeling, and a wide mouth design for easy addition and extraction of sample material. The externally-threaded design minimizes contamination risk in manual handling procedures. The tubes and rack are rated for storage down to vapor phase liquid nitrogen temperatures.
Informations légales
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries