Principaux documents
TMO3665-0050
Nalgene® economy graduated cylinder
PMP, volume 50 mL, case of 18 ea
Synonyme(s) :
PMP graduated cylinder, PMP graduated flask
18 EACH
234,00 €
234,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
18 EACH
234,00 €
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
234,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
PMP
Conditionnement
case of 18 ea
Fabricant/nom de marque
Nalgene 3665-0050
Capacité
50 mL
Volume
50 mL
Description générale
Dispense liquids easily with simple to read, molded-in graduations that won′t rub off. Affordably priced Thermo Scientific Nalgene Polypropylene Economy Graduated Cylinders have a generous pour spout and exhibit excellent chemical and break resistance.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries