Principaux documents
TMO362002-9050
Nalgene® diagnostic bottle without cap
natural high-density polyethylene, volume 15 mL, case of 2000 ea
Synonyme(s) :
PPCO bottles, small volume bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
natural high-density polyethylene
Conditionnement
case of 2000 ea
Fabricant/nom de marque
Nalgene 362002-9050
Capacité
15 mL
Volume
15 mL
Description générale
Safely store and ship light-sensitive diagnostic samples and valuable reagents in small volumes with Thermo Scientific Nalgene Opaque Amber HDPE Diagnostic Bottles, used in test kits and many small-volume applications. These opaque amber high-density polyethylene bottles are for use with light sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries