Principaux documents
TMO312002-9025
Nalgene® diagnostic bottle
natural high-density polyethylene, volume 8 mL, case of 2000 ea
Synonyme(s) :
PPCO bottles, small volume bottle with closure
2000 EACH
1 410,00 €
1 410,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
2000 EACH
1 410,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
1 410,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
natural high-density polyethylene
Conditionnement
case of 2000 ea
Fabricant/nom de marque
Nalgene 312002-9025
Capacité
8 mL
Volume
8 mL
Description générale
Sample, store and ship small volume, valuable reagents. Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Bottles include linerless polypropylene closures that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries