Principaux documents
TMO2401-0250
Nalgene® economy wash bottle
low-density polyethylene, volume 250 mL, unlabelled, case of 36 ea
Synonyme(s) :
LDPE flush bottle, LDPE wash bottle
36 EACH
241,00 €
241,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
36 EACH
241,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
241,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
low-density polyethylene
Conditionnement
case of 36 ea
Fabricant/nom de marque
Nalgene 2401-0250
Capacité
250 mL
Volume
250 mL
Catégories apparentées
Description générale
Prevent separation when squeezing Thermo Scientific Nalgene LDPE Economy Wash Bottles, featuring a tight-fitting draw tube and closure. These low density polyethylene bottles are designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. Cut back tip to increase flow.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries