Principaux documents
TMO2123-0010
Nalgene® bottles, style 2123
natural high-density polyethylene, volume 4000 mL, case of 6 ea
Synonyme(s) :
nalgene bottle, nalgene bottles
6 EACH
276,00 €
276,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
6 EACH
276,00 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
276,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
natural high-density polyethylene
polypropylene copolymer bottle (PPCO)
Description
Autoclavable
Conditionnement
case of 6 ea
Fabricant/nom de marque
Nalgene 2123-0010
Capacité
4000 mL
Volume
4000 mL
Description générale
This wide-mouth polypropylene copolymer bottle with closure is excellent for collecting, storing and transporting dry chemicals and environmental samples. Square, space saving design features molded-in handgrips for greater convenience and safety when pouring, autoclavable.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries