Principaux documents
T1668
Nalgene® centrifuge tubes, Oak Ridge Style 3118
nominal capacity 28 mL
Synonyme(s) :
High-Speed ​​Centrifuge Tubes, Nalgene tubes
10 EA
108,00 €
108,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
10 EA
108,00 €
About This Item
Code UNSPSC :
41121703
Nomenclature NACRES :
NB.11
108,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polypropylene screw closure
round bottom tube
screw top cap
transparent polycarbonate
Caractéristiques
RCF 50,000 × g
Conditionnement
pack of 10 ea
Fabricant/nom de marque
Nalgene® 3118-0028
Capacité nominale
28 mL
Description générale
“Nalgene” Oak Ridge high-speed polycarbonate centrifuge tubes offer outstanding mechanical strength, allowing for a wide range of applications. Additionally, their transparent nature facilitates easy observation of the contents in the tube.
Caractéristiques et avantages
  • Capable of withstanding temperatures ranging from -135°C to +135°C (-211°F to +275°F).
  • Designed with a round bottom to accommodate high-speed rotors.
  • Suitable for use in both refrigerated and non-refrigerated centrifuges.
  • Includes polypropylene linerless closures, which are easy to clean and provide leakproof protection under normal conditions up to 10,000 x g.
  • Autoclavable.
Autres remarques
  • Polycarbonate centrifuge tubes will have their lifespan reduced with repeated autoclaving.
  • It is essential to fill the tubes to a minimum of 80% of their total capacity to ensure proper performance.
  • To achieve leakproof performance exceeding 10,000 x g, utilize a sealing cap or sealing cap assembly.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries