Principaux documents
T0693
Pharmed L/S tubing
size 15
Synonyme(s) :
Saint Gobain Peristaltic Pump Tubing
25 FT
210,00 €
210,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
25 FT
210,00 €
About This Item
Code UNSPSC :
41105108
Nomenclature NACRES :
NB.51
210,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 25 feet
Fabricant/nom de marque
Tygon
D.I.
0.19 in. (4.8 mm)
Description générale
Tubing for MasterFlex Peristaltic Pumps, PharMed tubing has been specifically formulated for the MasterFlex pump system. Great for tissue and cell work- nontoxic and nonhemolytic. Long service life minimizes risk of fluid exposure; reduces tubing cost and pump downtime.
  • Opaque to UV and visible light to protect light-sensitive fluids
  • Heat sealable, bondable, and formable
  • Extremely low gas permeability
Informations légales
MasterFlex is a trademark of Cole-Parmer Instrument Co.