Principaux documents
QTLSD
Supelco
Erlich test reagent for LSD and psilocybin
About This Item
Code UNSPSC :
41116107
Application
Reagent for presumptive color assay of LSD and psilocybin.
Johns, S.H., et al.
Journal of Forensic Sciences, 24, 631-631 (1979)