Principaux documents
OH30073979
Ohaus® Pen pH Meter
Model ST10T-B
About This Item
Code UNSPSC :
41115603
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 30073979
Plage de mesure des TDS
0.0-1000.0 mg/L
Alimentation par piles
4 × 1.5 V, 200 hr life
Description générale
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
Application
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
Caractéristiques et avantages
Easily replaceable electrodes, automatic shut off feature
Informations légales
Ohaus is a registered trademark of Ohaus Corp.