Principaux documents
MTCP8051
MTC Bio Divided Reservoir
capacity 50 mL, non-sterile, pack of 100 ea (bulk bag)
Synonyme(s) :
cryo tube rack, reagent dish, reagent holder
About This Item
Code UNSPSC :
41121522
Nomenclature NACRES :
NB.12
Stérilité
non-sterile
Conditionnement
pack of 100 ea (bulk bag)
Fabricant/nom de marque
MTC Bio P8051
Capacité
50 mL
Description générale
Reservoir, divided, 50ml non-sterile, bulk pack, 100/cs
Caractéristiques et avantages
  • Graduated inner walls
  • Micro-trough U-shaped bottoms
  • Pouring spouts on all 4 corners
Informations légales
MTC is a trademark of MTC Bio