Principaux documents
MT30671569
Sigma-Aldrich
EasyPlace Electrode Arm
complete
1 EACH
267,00 €
267,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EACH
267,00 €
About This Item
Code UNSPSC :
23242102
Nomenclature NACRES :
NB.79
267,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Mettler-Toledo 30671569
Compatibilité
for use with Mettler-Toledo meters
Description générale
Precise Control
Perfectly vertical sensor movement makes it easy to insert the sensor into samples.
The right place
The EasyPlace contains dedicated positions for calibration, short- and long term sensor storage and measurement, simplifying everyday workflows.
Fits right in
The EasyPlace electrode arm fits together with the EasyMix magnetic stirrer and standard sachets, simplifying operation and saving space.