Principaux documents
HS14521A
Aluminium weighing boat
D × H 43 mm × 12 mm , small
1 EACH
30,90 €
30,90 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EACH
30,90 €
About This Item
Code UNSPSC :
41121812
Nomenclature NACRES :
NB.35
30,90 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Fabricant/nom de marque
Heathrow Scientific HS14521A
P × H
43 mm × 12 mm , small
Description générale
designed for general-purpose weighing, dispensing, storage, food processing, evaporating, or drying.
Boats also make good dust covers for open vessels.
  • All weigh boats are FDA/food-grade compliant
  • Disposable
  • Aluminum, Weighing Boats
Sturdy yet lightweight boats feature a smooth, flat bottom for sample handling and a flanged, crimped wall for rigidity. Finger tab handle can be etched for identification prior to weighing.
3 sizes
Packs of 100
Les clients ont également consulté
Slide 1 of 1