Principaux documents
HS120243
Microtube Rack
Microtube Rack
blue Nylon, pack of 4 racks, Holds 96 x 1.5-2 mL tubes
4 EA
67,40 €
67,40 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
4 EA
67,40 €
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.22
67,40 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
blue Nylon
Caractéristiques
Holds 96 x 1.5-2 mL tubes
Conditionnement
pack of 4 racks
Fabricant/nom de marque
Heathrow Scientific HS120243
Technique(s)
PCR: suitable
Diam. des puits
11.1 mm
Caractéristiques et avantages
  • made from reinforced nylon
  • superior chemical and heat resistance
  • handles provide easy and safe transportation
  • stackable two formats available for smaller and larger requirements
  • temperature range -40 to 140C
  • autoclavable
Questions
  1. Qual è il diametro dei pozzetti e quindi quello delle provette che possono alloggiare?
    1 answer
    1. The diameter of the well is 11.1 mm and can accommodate a 1.5 mL tube and a 2.0 mL tube. This product can hold up to 96 tubes. The dimensions are 8.3 x 5.2 x 2.1 in (21 x 13.1 x 5.4 cm).
      Helpful?
Reviews
Active Filters
  1. 1 Ratings-Only Review