GN782270
Greiner MASTERBLOCK® storage microplate
Deep Well, 1536 well, polypropylene, capacity 18 uL, conical (V) bottom, translucent
Synonyme(s) :
1536 well microplate, 1536 well plate, 1536 well storage microplate, cell culture plate
About This Item
Matériaux
polypropylene
Conditionnement
pack of 60 ea
Fabricant/nom de marque
Greiner 782270
Disponibilité
available only in China
Taille
1536 wells
Adéquation
suitable for (general storage, liquid handling; automated applications)