Principaux documents
GN781185
Greiner microplate
384 well, flat bottom, clear
Synonyme(s) :
384 well microplate, 384 well plate, cell culture plate
About This Item
Code UNSPSC :
41121800
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
flat bottom
Conditionnement
pack of 32 ea
Fabricant/nom de marque
Greiner 781185
Disponibilité
available only in China
Taille
384 wells
Adéquation
suitable for (cell culture)
Description générale
  • Greiner microplates are ideal for storing active agents, patient samples, DNA/RNA or stock cultures.
  • Polypropylene material can withstand a high temperature (-196 to +121°C) and are chemically resistant.
  • Nonpyrogenic plates are free of detectable DNase, RNase, human DNA.
  • Features alphanumeric well coding for identification.
  • Sealable with adhesive films and heat sealers for long-term and secure sealing.