Principaux documents
GN767001
Greiner Multiwell Strips
medium binding, round bottom (8 well), non-sterile
Synonyme(s) :
Greiner Multiwell Strips
About This Item
Code UNSPSC :
41121800
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Stérilité
non-sterile
Conditionnement
pack of 400 ea
Fabricant/nom de marque
Greiner 767001
Disponibilité
available only in China
Taille
8 well
Type de liaison
medium binding surface
Description générale
Both medium-binding and high binding are offered. High binding plates and strips are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding are suggested.