Principaux documents
GN710107
Greiner CELLSTAR® serological pipette
volume 2 mL, 1 / 100 mL graduations, green nozzle
Synonyme(s) :
pipette, serological pipette, Greiner serological pipettes
About This Item
Code UNSPSC :
41121500
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Stérilité
sterile
Conditionnement
pack of 1000 ea
Fabricant/nom de marque
Greiner 710107
Disponibilité
available only in China
Description générale
Greiner pipettes are precision extruded from the highest quality, optical-grade polystyrene. All volumes are identified by the standard international color code convention. Accuracy is maintained by high surface polish, both internally and externally, to reduce meniscus formation and ensure complete liquid expulsion with minimum carry-over.
Precision is aided by fine-line graduation printing providing accuracy of better than ±2%.
Caractéristiques et avantages
  • 1 mL, 2 mL and 5 mL sizes incorporate drawn tips while larger sizes feature molded tips to provide maximum accuracy
  • Packed individually, then double packed for extra protection
  • Designed to fit all standard pipetting devices
  • Cotton plugged for safety
  • Sterilized by γ-irradiation
  • DNase, RNase & human DNA free
Informations légales
CELLSTAR is a registered trademark of Greiner Bio-One GmbH