Principaux documents
GESE25010A75
GE SE250 mini-vertical unit for two slab gels
complete, gel size 10 cm × 8 cm, AC/DC input 500 V AC
About This Item
Code UNSPSC :
41105300
Nomenclature NACRES :
NB.22
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Cytiva Article No. 80-6147-45
Cytiva SE250-10A-,75
Taille du gel
10 cm × 8 cm
Catégories apparentées
Description générale
Runs two standard 10×8 cm mini-gels. The upper buffer chamber is also an efficient heat exchanger for active temperature control. The unit can run 12 tube gels with two tube gel adapter kits.