GE10600060
Amersham™ Hybond® LFP Western blotting membranes, PVDF
pore size 0.2 μm, sheet W × L 200 mm × 200 mm, pkg of 10 ea
372,00 €
About This Item
372,00 €
Produits recommandés
Conditionnement
pkg of 10 ea
Fabricant/nom de marque
Cytiva 10600060
Feuille, l × L
200 mm × 200 mm
Dimension de pores
0.2 μm pore size
Description générale
Caractéristiques et avantages
- Much lower background fluorescence than other commercially available PVDF membranes, resulting in higher sensitivity.
- Protein binding capacity over 200 µg/cm2.
- Compatible with a variety of blocking agents, transfer buffers, and total protein blot stains.
- Available in popular precut sizes for laboratory applications.
Stockage et stabilité