Principaux documents
F5426
Sigma-Aldrich
Carestream® electron microscopy film 4489
About This Item
Code UNSPSC :
41105300
Nomenclature NACRES :
MA.01
Taille de la feuille
6.5 cm × 9 cm
Température de stockage
2-8°C
Description générale
Fine-grain, blue-sensitive emulsion on 7-mil ESTAR base, notched for emulsion identification. Tear- and abrasion-resistant, dimensionally stable. Requires shorter pump-down time and curls less than Kodak electron image film SO-163.
Informations légales
Kodak is a registered trademark of Eastman Kodak Company