EP6731011015
Eppendorf® CellXpert® C170i CO2 Incubator
Oxygen control, inner door non-segmented, handle right side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
Synonyme(s) :
Heaters
About This Item
Code UNSPSC :
41104405
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 6731011015
Catégories apparentées
Description générale
CellXpert® C170i, non-segmented inner door, handle right side, 1 – 20 % oxygen control, 100 – 127 V/50 – 60 Hz (US/JP/South America)
Caractéristiques et avantages
- 1 – 20 % oxygen control
- handle right side
- non-segmented inner door
- 180° HTD (standard with C170i)
- including water tray and four shelves (stainless steel)
- 100 – 127 V, 50 – 60 Hz
Informations légales
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG