EP5804715403
Eppendorf® Aluminium-spare lid
non-aerosoltight, for rotor F-45-30-11
104,00 €
About This Item
104,00 €
Produits recommandés
Caractéristiques
non-aerosoltight, for rotor F-45-30-11
Fabricant/nom de marque
Eppendorf® 5804715403
Technique(s)
centrifugation: suitable
Compatibilité
for use with 5804 R
for use with 5804
for use with 5810 R
for use with 5810