EP5495512004
Rotor for Eppendorf® 5425 Centrifuge
S-96x0.2-PCR, to hold, 96 x 0.2 mL PCR tubes, to hold, 12 x 8-tube PCR strips, includes 4 x swing buckets
757,00 €
About This Item
757,00 €
Produits recommandés
Caractéristiques
to hold 12 x 8-tube PCR strips
to hold 96 x 0.2 mL PCR tubes
S-96x0.2-PCR
includes 4 x swing buckets
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5495512004
Paramètres
6,000 max. RCF (3,217 xg)
Technique(s)
PCR: suitable
centrifugation: suitable
Compatibilité
for use with centrifuge models 5425