Key Documents
EP5490034007
Rotor for Eppendorf® Concentrator Plus
F-45-72-8, non aerosol tight, to hold, 72 x 0.5 mL reaction vessels
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
to hold 72 x 0.5 mL reaction vessels
fixed angle rotor
non aerosol tight
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5490034007 (EU)
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models Concentrator Plus
Informations légales
Eppendorf is a registered trademark of Eppendorf AG