Principaux documents
EP5361000015
Eppendorf® Exchangeable Thermoblock
Holds 24 x 0.5 mL tubes, complete with IsoTherm Rack and IsoTherm cool pack 0 C
About This Item
Code UNSPSC :
41102404
Caractéristiques
Holds 24 x 0.5 mL tubes
complete with IsoTherm Rack and IsoTherm cool pack 0 C
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Eppendorf® 5361000015
Description générale
Product Information
For use with ThermoStat plus (heating/ cooling).
The exchangeable thermoblock system for the Eppendorf ThermoMixer® comfort and ThermoStat plus ensures a precise “fit” for all common lab tubes: from simple temperature control of a chemical to complex enzyme reactions and hybridization experiments, and from MTPs to 50 mL conical tubes – the application possibilities are virtually endless!
Features
  • A wide range of exchangeable thermoblocks to cover tubes from 0.2 mL to 50 mL as well as plates and glass slides
Heat insulated thermoblocks for ergonomic and safe operation (no direct contact with hot surfaces - you won′t burn your fingers)
  • Dry incubation technology eliminates contamination risk associated with classical waterbath incubation
  • Simple replacement of thermoblocks
  • Tubes are easy accessible
  • Whisper quiet operation and optimal temperature transfer due to complete enclosure and custom fit of tubes in the exchangeable thermoblocks
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Thermomixer is a registered trademark of Eppendorf AG