EP5242956003
Eppendorf® Microloader
pkg of 192 ea
Synonyme(s) :
Flow indicator
About This Item
Code UNSPSC :
41121600
Produits recommandés
Conditionnement
pkg of 192 ea
Fabricant/nom de marque
Eppendorf® 5242956003
Description générale
- Microloader™ tips are extremely long, fine, and flexible for filling microcapillaries for microinjection
- Unique tips are ideal for recovering surplus solution from the capillary
- Provides the ideal solution where additional reach is needed while pipetting the smallest volumes.
Caractéristiques et avantages
- Microloader tip with a volume range of 0.5-20 uL
- Light gray color tips of 100mm length
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Microloader is a trademark of Eppendorf AG